missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Streptavidin Magnetic Beads

Product Code. 17873044
Click to view available options
Product Type:
Dynabeads™ M-270 Streptavidin
Dynabeads™ M-280 Streptavidin
Dynabeads™ MyOne™ Streptavidin C1
Dynabeads™ MyOne™ Streptavidin T1
Dynabeads™ Streptavidin Trial Kit
Dynabeads™ Streptavidin for Target Enrichment
Isolation Kit, Dynabeads™ kilobaseBINDER™ Kit
Streptavidin Conjugate, Dynabeads™ MyOne™ Streptavidin T1
Quantity:
10 mL
100 mL
2 mL
200 Isolations
4 x 1 mL
50 mL
Unit Size:
10mL
2mL
50mL
Each
This item is not returnable. View return policy

Product Code. 17873044

Brand: Invitrogen™ 65607D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Supplied in PBS, pH 7.4 with 0.1% BSA and 0.02% NaN3 added as preservatives.

Dynabeads Streptavidin Magnetic Beads are designed for high-specificity, low-background capture of target molecules with biotinylated affinity binders such as antibodies, antigens, or oligonucleotides. Available in various sizes and surface chemistries, these beads can be tailored to provide the best fit for specific application needs.

Dynabeads Streptavidin Magnetic Beads are designed for high-specificity, low-background capture of target molecules with biotinylated affinity binders such as antibodies, antigens, or oligonucleotides. Available in various sizes and surface chemistries, these beads can be tailored to provide the best fit for specific application needs.

Dynabeads Streptavidin Magnetic Beads are compatible with high-throughput automation platforms, including KingFisher sample purification systems, ensuring seamless integration into new and existing workflows.

Key advantages of Dynabeads Streptavidin Magnetic Beads

  • ISO 13485–certified manufacturing—exceptional manufacturing control to ensure robust and consistent performance between production lots, meeting international quality standards for medical devices
  • Monosized, monodispersed magnetic beads—uniform particle size provides consistent and reproducible results across applications
  • Dual size options—available in 1-μm and 2.8-μm diameter specifications to accommodate diverse experimental requirements and application-specific needs
  • Multiple surface chemistry options—various surface functionalization chemistries available to optimize binding efficiency and compatibility for specific applications
  • Application-optimized design—size and surface chemistry combinations enable best-fit selection for targeted experimental protocols and assay requirements

Precision-engineered manufacturing delivers exceptional control over critical bead properties

Dynabeads Streptavidin products are based on magnetic polystyrene-based beads that are covalently coupled with recombinant streptavidin, creating a robust platform for target capture and isolation using biotinylated affinity binders. Dynabeads Streptavidin product manufacturing enables exceptional precision over three critical parameters: size distribution, magnetization properties, and surface chemical characteristics. This ensures consistent performance across batches and applications, providing the reliability necessary for reproducible experimental outcomes.

Optimized size selection enhances application-specific performance

The Dynabeads Streptavidin product line offers two distinct size options, each engineered to excel in specific experimental contexts. The 1-μm beads, available in the Dynabeads MyOne series, are ideally suited for applications requiring fast reaction kinetics, slow sedimentation rates, and high sensitivity-to-capacity ratios. These smaller beads provide enhanced surface area contact and reduced gravitational settling, making them particularly valuable for sensitive detection assays.

In contrast, the 2.8-μm beads, found in the Dynabeads M-270 and M-280 series, are perfect for applications demanding low background binding and reduced risk of aggregation in aggregation-prone applications. These larger beads offer even easier handling and faster magnetic separation compared to the Dynabeads MyOne series, facilitating streamlined workflow processing.

Tailored surface chemistry options address diverse experimental requirements

The Dynabeads Streptavidin product line includes two distinct surface chemistry configurations designed to optimize performance across varied experimental conditions. Hydrophobic surfaces are available in Dynabeads MyOne Streptavidin T1, Dynabeads Streptavidin for Target Enrichment, and Dynabeads M-280 Streptavidin products. These surfaces are BSA-blocked after streptavidin coupling making them ideal for high signal-to-noise applications such as immunoassays, target enrichment, and on-bead PCR for next-generation sequencing.

Alternatively, hydrophilic surfaces are featured in Dynabeads MyOne Streptavidin C1, Dynabeads Streptavidin for In Vitro Transcription, and Dynabeads M-270 Streptavidin products. These surfaces contain no BSA blocking and are particularly suitable for hydrophobic targets, harsh lysis conditions, sample preparation workflows for mass spectrometry, and applications demanding low or no nuclease activity, such as on-bead reverse transcription or in vitro transcription.

For researchers uncertain about which bead type best suits their application, a Dynabeads Streptavidin Trial Kit is available. It allows for side-by-side performance evaluation of different bead formulations under your specific experimental conditions, ensuring optimal selection for your research needs.

ISO 13485 certification ensures scalable quality and OEM support

Manufactured under ISO 13485 certification, Dynabeads Streptavidin products deliver outstanding batch-to-batch consistency. This robust quality assurance framework ensures that once customers develop an assay or workflow, they can rely on reproducible results across multiple experimental iterations. This consistency makes these versatile beads an efficient solution for applications spanning the entire spectrum from fundamental research to commercial manufacturing processes. For researchers developing commercial products or services, or those requiring larger bulk volumes, specialized support is available by emailing oemdynal@thermofisher.com.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Bead Type Magnetic polystyrene-based beads covalently coupled with recombinant Streptavidin
Concentration 10 mg/mL
Content And Storage Dynabeads Streptavidin for Target Enrichment is supplied in PBS, pH 7.4, with 0.1% BSA, 0.02% NaN3. Store at 2°C to 8°C.

Format Beads in Suspension
Isolation Technology Magnetic Bead
Diameter (Metric) 1 μm (CV < 5%)
For Use With (Equipment) KingFisher™ Sample Purification System, DynaMag™ magnets
For Use With (Application) Capture of targeted DNA sequences for Next-Generation Sequencing (NGS) and hybridization-based DNA assays
High-throughput Compatibility High-throughput Compatible
Product Line Dynabeads
Product Type Dynabeads™ Streptavidin for Target Enrichment
Quantity 50 mL
Shipping Condition Ambient Temperature
Binding Property 950-1500 pmoles free biotin/mg beads
Regulatory Status For Research Use Only
Certifications/Compliance ISO9001 and ISO13485
Surface Functionality Tosylactivated hydrophobic surface blocked with BSA
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.