missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Dynabeads™ Spot-On™ Slides

Product Code. 10577203 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 slides
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10577203 100 slides Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10577203 Supplier Applied Biosystems™ Supplier No. 74004
  • ONLINE EXCLUSIVE PRICE
    £369.56
    £337.43 / Pack of 1
    Save £32.13
    8% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dynabeads™ Spot-On™ Slides

Treated, single 9 mm well, glass slides for microscopic analysis of oocysts and cysts. This product is used in combination with the Dynabeads™ anti-Cryptosporidium and GC-Combo kits.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Material Glass
Type Microscope Slide
Well Count 1
Quantity 100 slides

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.