missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ SARS-CoV-2 Spike

Product Code. 17468082
Click to view available options
Volume (Metric):
3 mL
5 x 3 mL
Unit Size:
15mL
3mL
This item is not returnable. View return policy

Product Code. 17468082

Brand: Invitrogen™ 18100D

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Dynabeads™ SARS-CoV-2 Spike consists of superparamagnetic beads coupled with a trimeric spike protein. The spike protein is a patented recombinant trimeric structure combining the S1 subunit, including the receptor binding domain (RBD), and the S2 subunit found on the surface of the SARS-CoV-2 virus.

The Dynabeads SARS-CoV-2 Spike consists of superparamagnetic beads coupled with a trimeric spike protein. The spike protein is a patented recombinant trimeric structure combining the S1 subunit, including the receptor binding domain (RBD), and the S2 subunit found on the surface of the SARS-CoV-2 virus. The virus spike protein binds to human host cells via the angiotensin-converting enzyme 2 (ACE2) receptor to initiate virus attachment, as well as to antibodies produced in the body as a response to SARS-CoV-2 infection.

Dynabeads SARS-CoV-2 Spike features include:
• Fast SARS-CoV-2 antibody binding kinetics
• Dynabeads-facilitated automation (limited hands-on time)
• Also available in SARS-CoV-2 ELISA kits

This Dynabeads SARS-CoV-2 Spike is also included in our enzyme‐linked immunosorbent assay (ELISA) kits designed to detect and quantify the level of human SARS-CoV-2 IgG (add link to #18000D), IgM (add ink to #18010D), and Total Ig (add link to #18020D) antibodies from serum and plasma samples. These kits can be used manually with a DynaMag-96 magnet or in an automated fashion using a KingFisher instrument. The magnetic beads significantly improve the binding kinetics to cut the ELISA incubation time in half (45 minutes) compared to the more conventional plate-based ELISAs (90 minutes).

Since the spike protein binds host cells to initiate the entrance of SARS-CoV-2 virus into the epithelial cells of the lungs via ACE2, the beads may also be used for isolation of ACE2+ cells or for ACE2 protein purification.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 13485
Concentration 1 mg/mL
Content And Storage The beads are supplied in 0.05 M Tris-buffered saline (TBS) with 0.001% Tween-20 and 0.02% NaN3.

Store at 2°C to 8°C.

Detection Method Absorbance at 450 nm
Color Brown
For Use With (Application) Isolation of SARS-CoV-2 Cells
Material Magnetic Polystyrene
Product Type SARS-CoV-2 Spike Protein Magnetic Beads
Format 3 mL
For Use With (Equipment) KingFisher™ Flex or Apex Instruments, DynaMag™-96 Side Skirted, Magnetic 96-Well Separator, ELISA Plate Reader (450 nm)
High-throughput Compatibility Yes, on KingFisher™ Flex or Apex instruments
Isolation Technology Dynabeads™ magnetic beads
Shipping Condition Wet ice (2° to 8°C)
Volume (Metric) 3 mL
Diameter (Metric) 2.7 μm
Show More Show Less

For research use only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.