missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Protein G and Magnet Starter Pack

Product Code. 15716064
Change view
Click to view available options
Sufficient For:
100 reactions
40 reactions
Unit Size:
100 reactions
40 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
15716064 40 reactions 40 reactions
15781758 100 reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price
Product Code. 15716064 Supplier Invitrogen™ Supplier No. 10014D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase 2 of the same product at list price in a single order to receive 30% discount. There is no limit to the multiples of 2 that customers can buy. Use promo code ”27974” to get your promotional price

The Dynabeads Protein G and Magnet Starter Pack combines Dynabeads Protein G magnetic beads for immunoprecipitation with the magnet preferred for use with 1.5 mL microcentrifuge tubes for purchasing convenience.

Space-saving design with our smallest thermal cycler footprint

  • Flexible modular design saves space on deck
  • Separate control panel module provides flexibility when integrating on the robotic deck
  • Choice of 10 cm,1 meter and 3 meter cabling options

 

For Research Use Only. Not for use in diagnostic procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The product contains:
• 2 x 1 mL Dynabeads Protein G beads
• 1 x DynaMag-2 magnet

Store Dynabeads at 2–8°C.
Shipping Condition Room Temperature
Product Line DynaMag, Dynabeads
Quantity 40 reactions
Type Magnet Starter Pack
Ligand Type Protein G
Sufficient For 40 reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.