Learn More
Invitrogen™ Dynabeads™ Protein G IP Kit and Magnet Starter Pack
Description
The Dynabeads Protein G IP Kit and Magnet Starter Pack combines Dynabeads Protein G magnetic beads for immunoprecipitation (including the required buffers) and the magnet preferred for use with 1.5 mL microcentrifuge tubes for purchasing convenience. The beads and buffers are sufficient for 40 reactions, and the magnet holds 16 x 1.5 mL microcentrifuge tubes. The Immunoprecipitation Kit - Dynabeads Protein G with buffers, stand-alone Dynabeads Protein G beads, and the DynaMag-2 magnet are also available separately.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
| Kit Contents | 2 mL Dynabeads Protein G beads; 16 mL Antibody Binding and Washing Buffer; 28 mL Washing Buffer; 1 mL Elution Buffer; 1 x DynaMag-2 Magnet |
| Content And Storage | Store Dynabeads at 2–8°C. |
| Shipping Condition | Room Temperature |
| Product Line | DynaMag™, Dynabeads™ |
| Quantity | 40 reactions |
| For Use With (Application) | Immunoprecipitation |
| Type | IP Kit and Magnet Starter Pack |
| Ligand Type | Protein G |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.