missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Protein A/Protein G and Magnet Starter Pack

Product Code. 15700817
Change view
Click to view available options
Quantity:
40 reactions
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15700817 40 reactions 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15700817 Supplier Invitrogen™ Supplier No. 10015D
  • ONLINE EXCLUSIVE PRICE
    £660.00
    £613.65 / 1 sets
    Save £46.35
    7% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Dynabeads Protein A/Protein G and Magnet Starter Pack combines Dynabeads Protein A and Dynabeads Protein G magnetic beads for immunoprecipitation with the magnet preferred for use with 1.5 mL microcentrifuge tubes for purchasing convenience.

Space-saving design with our smallest thermal cycler footprint

  • Flexible modular design saves space on deck
  • The separate control panel module provides flexibility when integrating on the robotic deck
  • Choice of 10 cm, 1 meter, and 3 meter cabling options

Automated lid and cycler features

  • 384-well block
  • The automated lid is compatible with programming for easy hands-free operation with liquid handling or plate stackers
  • The heated cover slides forward to cover the plate nest,ensuring thermal contact and eliminating evaporation

For Research Use Only. Not for use in diagnostic procedures.

TRUSTED_SUSTAINABILITY

Specifications

Kit Contents 1 mL Dynabeads Protein A beads; 1 mL Dynabeads Protein G beads; 1 x DynaMag-2 magnet
Content And Storage Store Dynabeads at 2–8°C.
Shipping Condition Room Temperature
Product Line DynaMag, Dynabeads
Quantity 40 reactions
For Use With (Application) Immunoprecipitation
Type Magnet Starter Pack
Ligand Type Protein A/G

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.