missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Dynabeads™ MX Mixer Base

Product Code. 10272963 Shop All Applied Biosystems Products
Click to view available options
For Use With (Equipment):
Dynabeads™ Sample Mixers
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10272963

Brand: Applied Biosystems™ 15902

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Dynabeads™ MX Mixer Base

Dynabeads™ MX mixers provide continuous gentle motion to obtain optimal binding of Dynabeads™ with targets (e.g., cells, proteins and nucleic acids). Variable speed (5-40 rpm). Can be operated in both cold and warm conditions (4-37°C). Inter-changeable heads and a turret unit are designed to attach to the mixer base and enable usage with different tube sizes and sample volumes.
TRUSTED_SUSTAINABILITY

Specifications

Type MX Mixer Base
Content And Storage Store at room temperature.
For Use With (Equipment) Dynabeads™ Sample Mixers
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.