Learn More
Applied Biosystems™ Dynabeads™ MX 12-tube Mixing Wheel
Product Code. 10364393 Shop All Applied Biosystems Products
Change view
Click to view available options
For Use With (Equipment):
Dynabeads™ Sample Mixers
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10364393 Dynabeads™ Sample Mixers Each
1 options
Product Code. 10364393 Supplier Applied Biosystems™ Supplier No. 15903
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
Dynabeads™ MX 12-tube Mixing Wheel
Attachment for Dynabeads™ MX Mixer Base (Cat. No. 159-02). Designed to hold 12 sample tubes ranging from 1.5 ml (microcentrifuge) to 1.7 mm diameter. Requires use of Mixer Turret (Cat. No. 159-06) to affix to Mixer Base.
TRUSTED_SUSTAINABILITY
Specifications
Tube Capacity 1.5 mL
Product Type Mixing Wheel
For Use With (Equipment) Dynabeads™ Sample Mixers
No. of Positions 12