Learn More
Applied Biosystems™ Dynabeads™ L10 Tubes
Product Code. 10139012
Change view
Click to view available options
Quantity:
5 Tubes
Unit Size:
Pack of 5
Product Code. Quantity unitSize
10139012 5 Tubes Pack of 5
1 options
Product Code. 10139012 Supplier Applied Biosystems™ Supplier No. 74003
Dynabeads L10 Tubes
Flat-sided reusable glass tubes necessary for optimal recovery of cysts/oocysts during magnetic separation with the MPC-1 or MPC-6 magnetic particle concentrators when using Dynabeads™ anti-Cryptosporidium or Dynabeads™ GC-Combo.
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
For Use With Dynabeads™ MPC™
Quantity 5 Tubes
Product Type Collection Tube