missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Dynabeads™ Human T-Expander CD3/CD28

Product Code. 17755274
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 17755274

Brand: Gibco™ 11141D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dynabeads™ Human T-Expander CD3/CD28

Dynabeads™ Human T-Expander CD3/CD28 is intended for separation, activation and expansion of human T cells. Dynabeads™ Human T-Expander CD3/CD28 is the research grade version of Dynabeads™ CD3/CD28 CTS™. Containing the same amount and ratio of antibodies from the same clones as Dynabeads™ CD3/CD28 CTS™, Dynabeads™ Human T-Expander CD3/CD28 are intended for small scale pre-clinical research. Dynabeads™ Human T-Expander CD3/CD28 offers a simple method for activation and expansion of T-cells that does not require antigen presenting cells or antigen. By combining anti-CD3 and anti-CD28 antibodies on Dynabeads™, the beads provide both the primary and co-stimulatory signals that are required for activation and expansion of T cells.

Applications:
Dynabeads™ Human T-Expander CD3/CD28 is the research grade version of Dynabeads™ CD3/CD28 CTS™. The Dynabeads™ CD3/CD28 CTS™ technology has been successfully used in ex vivo separation, activation and expansion of T-cells in a number of studies, including studies on HIV infection and cancer. Other potential applications for this novel technology are treatment of autoimmune and infectious diseases and prevention of complications associated with chemotherapy and allogeneic transplantation.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (Whole Population)
Clonality Monoclonal
Content And Storage This Product Contains: Dynabeads™ CD3⁄CD28 is supplied as a suspension containing 1 x 108 beads⁄ml in phosphate buffered saline (PBS), pH 7.4, wwith 0.1% human serum albumin (HSA).
Store at: 2-8°C
Product Type Antibody Coated Bead
Final Product Type Cells
Product Line DYNAL™, Dynabeads™
Purity or Quality Grade Research Grade
Quantity 10 mL
Reactivity Human
Sample Type Cell Cultures, PBMC
Shipping Condition Room Temperature
Target Species Human
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.