missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Dynabeads™ EPEC/VTEC O26

Product Code. 10404513 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10404513 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10404513 Supplier Applied Biosystems™ Supplier No. 71013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Dynabeads EPEC/VTEC O26

For the rapid selective concentration of E.coli 026. Method can be performed manually or can be automated by using a BeadRetriever™.

This Product Contains: Dynabeads™ coated with anti-E.coli O26 antibodies

Sensitivity: Sensitivity is very high and enables detection of 1 viable organism in 25g of sample or as few as 100 organisms / ml after pre enrichment
TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 mL
Target Species E. coli
Content And Storage Store in refrigerator (2–8°C).
For Use With (Equipment) Dynabeads™ MPC™, KingFisher mL Food Protection Purification System
Shipping Condition Room Temperature
Product Line Dynabeads

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.