missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
For the rapid selective concentration of E.coli O145. Method can be performed manually or can be automated by using a BeadRetriever™.
This Product Contains: Dynabeads™ coated with anti-E.coli O145 antibodies
Sensitivity: Sensitivity is very high and enables detection of 1 viable organism in 25g of sample or as few as 100 organisms / ml after pre enrichment
This Product Contains: Dynabeads™ coated with anti-E.coli O145 antibodies
Sensitivity: Sensitivity is very high and enables detection of 1 viable organism in 25g of sample or as few as 100 organisms / ml after pre enrichment
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Dynabeads™ EPEC/VTEC O145, 2mL, Suspension, E. coli O145 Genotype, 7.4 pH, Sensitivity is Very High and Enables Detection of 1 Viable Organism in 25g of Sample or as Few as 100 Organisms/mL After Pre Enrichment |
| For Use With (Application) | Food and Beverage, Industrial and Applied Science |
| Format | Suspension |
| For Use With (Equipment) | Dynabeads™ MPC™, BeadRetriever™ system |
| Includes | Dynabeads™ coated with anti-E.coli O145 antibodies |
| Quantity | 2 mL |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?