missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Dynabeads™ EPEC/VTEC O103

Product Code. 10413293 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
Each
Product Code. Quantity unitSize
10413293 2 mL Each
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10413293

Brand: Applied Biosystems™ 71011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Dynabeads EPEC/VTEC O103

For the rapid selective concentration of E.coli 0103. Method can be performed manually or can be automated by using a BeadRetriever™.

This Product Contains: Dynabeads™ coated with anti-E.coli 0103 antibodies

Sensitivity: Sensitivity is very high and enables detection of 1 viable organism in 25g of sample or as few as 100 organisms / ml after pre enrichment
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C).
Product Type Antibody Coated Bead
For Use With (Equipment) Dynabeads™ MPC™, KingFisher mL Food Protection Purification System
Prep Scale Small Volume Microbial Samples
Product Line Dynabeads
Quantity 2 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.