missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ CD3

Product Code. 10725693
Change view
Click to view available options
Quantity:
2 mL
5 mL
Unit Size:
2mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10725693 5 mL 5mL
30275450 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10725693 Supplier Invitrogen™ Supplier No. 11151D
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Dynabeads CD3 are superparamagnetic beads coupled with an anti-human CD3 antibody that enable easy isolation or depletion of human CD3+ T cells directly from any sample, including whole blood, bone marrow, buffy coat, PBMC, and tissue digests.

Dynabeads CD3 are superparamagnetic beads coupled with an anti-human CD3 antibody that enable easy isolation or depletion of human CD3+ T cells directly from any sample, including whole blood, bone marrow, buffy coat, peripheral blood mononuclear cells (PBMCs), and tissue digests. Advantages of Dynabeads CD3:

  • Fast and efficient isolation of CD3+ T cells from any sample—no columns required
  • High purity, yield, and viability of positively isolated CD3+ T cells for molecular downstream assays
  • Fast and efficient depletion of CD3+ T cells from any sample.

About Dynabeads CD3

Dynabeads CD3 are uniform, superparamagnetic beads (4.5 μm diameter) coated with a primary monoclonal antibody specific for the CD3 membrane antigen mainly expressed mainly on human mature T cells. Because of the bead size, Dynabeads CD3 can easily and efficiently isolate or deplete cells from viscous samples such as whole blood and bone marrow in about 30 minutes. Dynabeads CD3 will also afford excellent recovery of high-purity, viable cells for use in downstream molecular studies; for example, those in which cells are to be lysed while still attached to beads, and nucleic acids or proteins further purified. Note that intact cells will not be released from these beads, so if you want to isolate CD3+ cells for cell-based applications, or you need to check your samples with flow cytometry, check out our total human T cell product offerings.

Magnetic bead to based separation offers easy handling

Dynabeads CD3 is added to the sample under continuous mixing to optimize the binding of the Dynabeads to the target cells. By placing the sample on a magnet, the bead-bound target cells are separated from the rest of the sample in just 1 to 2 minutes. For depletion, remove the supernatant to a new tube for further studies and discard the bead-bound cells. For positive isolation for molecular studies, remove the supernatant and wash the bead-bound cells 2 to 3 times in buffer to get optimal purity. The cells can be lysed while still attached to the beads and the supernatant transferred to a new tube for downstream molecular analysis. Starting samples can be whole blood, buffy coat, bone marrow, PBMC, or tissue digests.

Automation-ready for KingFisher purification systems

The product is ideal for high-throughput workflows using KingFisher systems. A user guide and scripts for KingFisher Flex and Apex workflows are available for download on this page.

Learn more about Dynabeads products

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (Whole Population)
Content And Storage This Product Contains: 5 mL Dynabeads™ CD3 coated with anti-CD3 monoclonal antibody.

Storage Conditions: 2°C to 8°C. Do not freeze.
Product Type Antibody Coated Bead
Includes 5 mL Dynabeads CD3 coated with anti-CD3 monoclonal antibody.
Isolation Technology Depletion, positive isolation for molecular applications
No. of Cells Processes ∼2 x 109 cells total
Output Viability >95%
Product Line DYNAL, Dynabeads
Purity or Quality Grade Research Grade
Quantity 5 mL
Sample Type PBMC, Blood
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 PBMCs per isolation
Target Species Human
Diameter (Metric) 4.5 μm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.