missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dynabeads™ CaptureSelect™ AAVX Magnetic Beads

Product Code. 30276684 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
50 mL
Unit Size:
1mL
50mL
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30276684 5 mL Each
30276387 1 mL 1mL
30276198 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30276684 Supplier Thermo Scientific™ Supplier No. 2853522005
  • ONLINE EXCLUSIVE PRICE
    £1460.00
    £1258.00 / Pack of 1
    Save £202.00
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Dynabeads™ CaptureSelect™ AAVX Magnetic Beads are high-capacity, high-throughput magnetic affinity particles for the purification of a broad range of naturally occurring and synthetic adeno-associated virus (AAV) serotypes used for gene therapy applications.

Dynabeads CaptureSelect AAVX Magnetic Beads are high-capacity, high-throughput magnetic affinity particles for the purification of a broad range of naturally occurring and synthetic adeno-associated virus (AAV) serotypes used for gene therapy applications. The anti-AAVX affinity ligand demonstrates good binding reactivity towards a large variety of AAV serotypes, including AAV1 to AAV8, AAVrh10, and synthetic serotypes. The magnetic beads enable purification using manual and robotic magnetic separators of AAV serotypes directly from crude feedstock in a single step with high purity and yield.

Features of DynaBeads CaptureSelect AAVX Magnetic Beads include:

  • One-magnetic-step AAV purification from crude material with high purity and yield
  • Compatible with manual and automated workflows
  • High specificity and capacity
  • Easy upscaling to higher volumes
  • Non-animal derived

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a ∼12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Bead matrix: superparamagnetic
  • Bead size: 4.5 μm
  • Ligand: CaptureSelect AAVX affinity ligand
  • Binding capacity: ∼1 x1012 viral particles/mg beads
  • Elution conditions: 50 mM citric acid pH 2.5-3.0
  • Formulation buffer: tris-buffered saline, 0.05% Tween 20, 0.05 % sodium azide

Specifications

Concentration 30 mg beads/mL slurry, 1 mg contains approximately 1.3 x 107 beads
Formulation 4 to 6% (v/v) slurry in Tris buffered saline with 0.05% Tween 20 and 0.05% sodium azide
Matrix Non-porous monodisperse superparamagnetic beads
Content And Storage Store at 2°C to 8°C.
Shipping Condition Wet Ice
Particle Size 4.5 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 5 mL
For Use With (Application) Magnetic isolation of AAV capsids
Target Multiple AAV serotypes
Type Magnetic Affinity Bead
Ligand Type AAVX Affinity Ligand
Sufficient For Sample Preparation
Binding Capacity ∼1 x 1012 Viral Particles/mg Beads
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.