missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Dynabeads™ anti-E. coli O157

Product Code. 11593367 Shop All Applied Biosystems Products
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
5mL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11593367

Brand: Applied Biosystems™ 71004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Dynabeads E. coli anti-O157 is a tool for better sample preparation and improved cell culture and immuno-based assays for E. coli O157:H7 in food, feces, water and other environmental samples.

Dynabeads E. coli anti-O157 is a tool for better sample preparation and improved cell culture and immuno-based assays for E. coli O157:H7 in food, feces, water and other environmental samples. Based on well-known and widely used immunomagnetic separation (IMS) technology, Dynabeads E. coli anti-O157 provides a useful means to selectively capture E. coli O157:H7 and better prepare samples for effective analysis.

With Dynabeads anti-E. coli O157, you will:

  • Isolate and concentrate E. coli O157:H7 from samples in as little as 24 hours
  • Improve detection sensitivity in cell culture and immunoassays of samples
  • Perform either manual or automated (using the Thermo Scientific KingFisher mL Food Protection Purification System) separations
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Each unit of product contains 5 mL Dynabeads™ anti-E. coli O157.
Store at 2°C to 8°C.
Product Type Anti-E.coli O157 Bead
For Use With (Equipment) Dynabeads™ MPC™, KingFisher mL Food Protection Purification System, DynaMag™-2
Isolation Technology Immunomagnetic Separation
Prep Scale Small Volume Microbial Samples
Product Line Dynabeads
Quantity 5 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.