missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYM Monoclonal Antibody (1C2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA522064
This item is not returnable.
View return policy
Description
Peptide Sequence: QTSDQATLLL YTLLHQNSNI RTYMLARTDM ENLVLPILEI LYHVEERNSH HVYMALIILL ILTEDDGFNR SIHEVILKNI TWYSERVL
This gene encodes a protein which is necessary for normal skeletal development and brain function. Mutations in this gene are associated with two types of recessive osteochondrodysplasia, Dyggve-Melchior-Clausen (DMC) dysplasia and Smith-McCort (SMC) dysplasia, which involve both skeletal defects and mental retardation.
Specifications
| DYM | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q7RTS9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54808 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| 1C2 | |
| Unconjugated | |
| DYM | |
| DYM | |
| DYM (NP_060123, 343 a.a. ∽ 430 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction