missing translation for 'onlineSavingsMsg'
Learn More

DYKDDDDK Epitope Tag Antibody (L5), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18254164 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18254164 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18254164 Supplier Novus Biologicals Supplier No. NBP106712AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

DYKDDDDK Epitope Tag Monoclonal antibody specifically detects DYKDDDDK Epitope Tag in Epitope Tag samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DYKDDDDK Epitope Tag
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone L5
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DDDDK epitope tag, DDK, DYKDDDDK tag, ECS epitope tag, ECS tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, epitope tag flag, FLAG (FLAG is a trademark of Sigma Aldrich), Flag tag, Ty 1 Tag, xxxDDDDK epitope tag
Host Species Rat
Immunogen DYKDDDDK Epitope Tag Antibody (L5) was made to N-terminal DYKDDDDK-tagged extracellular domain of mouse Langerin. Binds to same epitope as SigmA′s Anti-FLAG® M2 Antibody.
Molecular Weight of Antigen 1.01 kDa
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Binds to same epitope as Sigma's Anti-FLAGM2 Antibody. FLAG is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
Target Species Guinea Pig
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.