missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Biodegradable and nontoxic.
- Blue dye brightens ponds.
- Dyes diffuse rapidly in water, producing a vivid, easy-to-see color.
- Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more.
- Use blue dye to brighten murky ponds and other landscaped waters.
- Minimum detectable concentration of all dyes in clear water is 1 ppm.
- Available in tablets, liquid, or powder.
- Quick dissolving tablets are easy to handling with less mess than other forms.
- They are perfect for applications such as storm lines, cooling and plumbing systems, and sewer lines.
- Choose liquid for applications such as metered input, flow/dilution/disperation rate assessment, septic inspections, or fluoremetric dye tests.
- Powder is well-suited for large applications where liquid is undesirable such a cooling towers, lakes, rivers, and ponds, or power plant piping.
Specifications
Specifications
| Color | Blue |
| Description | Blue Dye Tablets, 200 Tablets |
| For Use With (Application) | Brightening murky ponds and other landscaped waters |
| No. of Tests | 200 Tests |
| Quantity | 200 Pk. |
| Sterility | Non-sterile |
| Type | Dye Tracer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?