missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Dye Concentrate
Shop All Cole-Parmer Products
Click to view available options
Color:
Blue
Dark Red
Yellow/Green
pH Range:
5.1 to 5.3
8
8.6 to 8.8
Unit Size:
Each
Description
F
- Fluorescent yellow/green dye (fluorescein) works best in silt-filled waters.
- Use an ultraviolet lamp (order separately below) to trace fluorescent red or yellow/green dyes where normal visual detection is difficult
- Won’t harm the environment—nontoxic and biodegradable
- Easy-to-use color in rapidly diffusing dye identifies dispersion patterns, flow rates, seepage and more Noncarcinogenic fluorescent red dye is ideal for yellowed or heavily algaed water
- Brighten murky ponds with blue dye
- Minimum detectable concentration is 1 ppm
Specifications
Specifications
| Color | Blue |
| pH Range | 5.1 to 5.3 |
| For Use With (Application) | Squirt-top dispensing |
| Quantity | 1/Pk. |
| Type | Dye Tracer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction