missing translation for 'onlineSavingsMsg'
Learn More

Dydrogesterone, MedChemExpress

Product Code. 16454317
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16454317

Brand: Medchem Express HYB0257A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dydrogesterone is a potent, orally active progestogen indicated in a wide variety of gynaecological conditions related to progesterone deficiency.

Dydrogesterone is a potent, orally active progestogen indicated in a wide variety of gynaecological conditions related to progesterone deficiency.

Specifications

Chemical Name or Material Dydrogesterone
Physical Form Powder
CAS 152-62-5
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C21H28O2
Solubility Information DMSO : 33.33 mg/mL (106.67 mM; Need ultrasonic)
SMILES CC([C@H]1CC[C@@]2([H])[C@]3([H])C=CC4=CC(CC[C@@]4(C)[C@]3([H])CC[C@]12C)=O)=O
Molecular Weight (g/mol) 312.45
Formula Weight 312.45
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.