missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
DYDC2 Monoclonal specifically detects DYDC2 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | DYDC2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 9 |
| Conjugate | Biotin |
| Dilution | ELISA |
| Formulation | PBS |
| Gene Alias | bA36D19.6, DPY30 domain containing 2, DPY30 domain-containing protein 2, MGC16186 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human DYDC2 (Q96IM9; Thr102-Pro176). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?