missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYDC1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-47218
This item is not returnable.
View return policy
Description
DYDC1 Polyclonal specifically detects DYDC1 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| DYDC1 | |
| Polyclonal | |
| Western Blot 0.5 - 1 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_620167.1 | |
| DYDC1 | |
| Peptide with sequence C-RNEDILHSEEATLD corresponding to internal region according to NP_620167.1. | |
| 0.1 mg | |
| Primary | |
| This antibody recognizes and epitope within the Internal region. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| BC019250 protein, DPY30 domain containing 1, DPY30 domain-containing protein 1, DPY30D1bA36D19.5, FLJ43920 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 143241 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction