missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DY131

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Novel selective agonist at estrogen-related receptors ERRβ and ERRγ. Displays minimal activity at ERRα, ERα and ERβ at concentrations up to 30 μM.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 311.38 |
| Purity | >99% |
| Molecular Weight (g/mol) | 311.385 |
| Chemical Name or Material | DY131 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction