missing translation for 'onlineSavingsMsg'
Learn More

DY 131, Thermo Scientific™

Product Code. 15466769
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15466769 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15466769 Supplier Thermo Scientific Alfa Aesar Supplier No. J65171.MA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A selective agonist at estrogen-related receptors ERR→ and ERR→

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 95167-41-2
Color Yellow
Molecular Formula C18H21N3O2
MDL Number MFCD00715585
Quantity 10 mg
Synonym unii-5vwv92039e, n-4-diethylaminobenzylidenyl-n'-4-hydroxybenzoyl-hydrazine, d06jpq, n'-4-diethylamino phenyl methylidene-4-hydroxybenzohydrazide, n-e-4-diethylamino phenyl methylideneamino-4-hydroxybenzamide, benzoic acid, p-hydroxy-, p-diethylamino benzylidene hydrazide, n'-1e-4-diethylamino phenyl methylene-4-hydroxybenzohydrazide, benzoic acid, 4-hydroxy-, 4-diethylamino phenyl methylene hydrazide, benzoic acid, 4-hydroxy-, 2-4-diethylamino phenyl methylene hydrazide, dy hplc
InChI Key WLKOCYWYAWBGKY-CPNJWEJPSA-N
SMILES CCN(CC)C1=CC=C(C=C1)C=NNC(=O)C2=CC=C(C=C2)O
IUPAC Name N-[(E)-[4-(diethylamino)phenyl]methylideneamino]-4-hydroxybenzamide
Molecular Weight (g/mol) 311.385
PubChem CID 5497124
Formula Weight 311.38
Physical Form Solid
Chemical Name or Material DY 131
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.