missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DXR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144466
This item is not returnable.
View return policy
Description
DXR Polyclonal Antibody for Western Blot, ELISA
Catalyzes the NADPH-dependent rearrangement and reduction of 1-deoxy-D-xylulose-5-phosphate (DXP) to 2-C-methyl-D-erythritol 4-phosphate (MEP).
Specifications
| DXR | |
| Polyclonal | |
| Unconjugated | |
| dxr | |
| ispC; yaeM | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 75202014 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 4.28 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P45568 | |
| dxr | |
| Recombinant Escherichia coli (strain K12) DXR protein. | |
| 100 μL | |
| Primary | |
| Bacteria | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction