missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DVL2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12037-1-AP-150UL
This item is not returnable.
View return policy
Description
Disheveled DVL2 is a member of the dishevelled (dsh) protein family. The vertebrate dsh proteins have approximately 40% amino acid sequence similarity with Drosophila dsh. This gene encodes a 90-kD protein that undergoes posttranslational phosphorylation to form a 95-kD cytoplasmic protein, which may play a role in the signal transduction pathway mediated by multiple Wnt proteins. The mechanisms of dishevelled function in Wnt signaling are likely to be conserved among metazoans.Specifications
| DVL2 | |
| Polyclonal | |
| Unconjugated | |
| DVL2 | |
| Dishevelled 2, DSH homolog 2, DVL2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13543, 1856 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O14641, Q60838 | |
| DVL2 | |
| DVL2 Fusion Protein Ag2666 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction