Learn More
DVL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27384-1-AP-20UL
 View more versions of this product
Product Code. 16805605
  • £135.00 / 20µL
Description
DVL1, the human homolog of the Drosophila dishevelled gene (dsh) encodes a cytoplasmic phosphoprotein that regulates cellproliferation, acting as a transducer molecule for developmentalprocesses, including segmentation and neuroblast specification.DVL1 is a candidate gene for neuroblastomatous transformation. TheSchwartz-Jampel syndrome and Charcot-Marie-Tooth disease type 2Ahave been mapped to the same region as DVL1. The phenotypes ofthese diseases may be consistent with defects which might beexpected from aberrant expression of a DVL gene during development.
Specifications
Specifications
DVL1
Polyclonal
Unconjugated
DVL1
Dishevelled 1, DSH homolog 1, DVL, DVL1
Rabbit
Antigen Affinity Chromatography
RUO
13542, 1855
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O14640, P51141
DVL1
DVL1 Fusion Protein Ag26055
20 μL
Primary
Human, Mouse