missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DVL1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67672-1-IG-150UL
This item is not returnable.
View return policy
Description
The DVL1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DVL1. This antibody recognizes human, rat, mouse antigen. The DVL1 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| DVL1 | |
| Monoclonal | |
| 1100 μg/mL | |
| DVL; MGC54245 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Rat, Mouse | |
| 70-75 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1A2A5 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 1855 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur