missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ DVB/Carboxyl Latex Beads, 4% w/v, 1μm
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
Description
Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Because of the way that the polystyrene chains arrange themselves in the bead, the surface is very hydrophobic in character, making these ideal materials for the adsorption of materials such as proteins.
The DVB/carboxyl microspheres are negatively charged cross-linked polystyrene microspheres with carboxyl functional groups on the surface. The surface charge is pH dependent and hydrophobic in nature. These beads are stable at neutral to high pH.
Microspheres Are Available in Many Sizes and Surface Functionalities
25 sizes to choose from—0.02 microns to 16 microns
20 different surface—for ready conjugation to virtually any material
Surfactant free—ultraclean
Various quantities—from milliliter to 500-liter amounts
Key Applications of Microspheres
Instrument calibration (flow cytometry, microscopy, HTS, HCS)
Flow testing (microfluidics, blood flow, water flow, and air flow)
Cell biology tracers (cell differentiation and cell tracing)
Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)
Get More Information
Read a technical overview of microspheres and find out about high activity beads, super active beads, specialty beads, and review bead-coupling protocols on the IDC Surfactant-free Latex Beads application page.
We’ll Make a Custom Microsphere Product for You
If you don’t see the microsphere you’re looking for in our stocked list, we’ll make a custom microsphere for you. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 9001:2000 certified.
For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
The DVB/carboxyl microspheres are negatively charged cross-linked polystyrene microspheres with carboxyl functional groups on the surface. The surface charge is pH dependent and hydrophobic in nature. These beads are stable at neutral to high pH.
Microspheres Are Available in Many Sizes and Surface Functionalities
Key Applications of Microspheres
Get More Information
Read a technical overview of microspheres and find out about high activity beads, super active beads, specialty beads, and review bead-coupling protocols on the IDC Surfactant-free Latex Beads application page.
We’ll Make a Custom Microsphere Product for You
If you don’t see the microsphere you’re looking for in our stocked list, we’ll make a custom microsphere for you. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 9001:2000 certified.
For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
Specifications
Specifications
| Concentration | 0.04w/v |
| Content And Storage | 2°C to 8°C |
| Description | DVB/Carboxyl Latex Beads, 15mL, 4% w/v Concentration, 1μm Particle Size, White, Suspension Form, De-ionized Water Medium, Specialty Latex Bead, Room Temperature, 2 to 8°C Storage Temperature, Ideal Materials for the Adsorption of Materials Such |
| Color | White |
| For Use With (Application) | Instrument Calibration (Flow Cytometry, Microscopy, HTS, HCS), Flow Testing (Microfluidics, Blood Flow, Water Flow, and Air Flow), Cell Biology Tracers (Cell Differentiation and Cell Tracing), Immunoassays (Agglutination Tests, ELISA, Particle Capture, and Contrast Reagents) |
| Material | Polystyrene |
| Format | Suspension |
| Packaging | 1 Each |
| Parking Area | 152 A2/COOH (carboxyl group) |
| Particle Density | 1.055g/cm3 at 20°C |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction