missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUX5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575648
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The human genome contains hundreds of repeats of the 3.3-kb family in regions associated with heterochromatin. The DUX gene family, including DUX5, resides within these 3.3-kb repeated elements (Beckers et al., 2001 [PubMed 11245978]). See DUX4 (MIM 606009).[supplied by OMIM, Mar 2008].
Specifications
| DUX5 | |
| Polyclonal | |
| Unconjugated | |
| DUX5 | |
| double homeobox 5; double homeobox protein 1; double homeobox protein 5; double homeobox, 5; DUX1; DUX5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26581 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96PT3 | |
| DUX5 | |
| Synthetic peptide corresponding to residues in Human DUX5. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction