missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUX5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£378.00
Specifications
| Antigen | DUX5 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
DUX5 Polyclonal specifically detects DUX5 in Human samples. It is validated for Western Blot.Specifications
| DUX5 | |
| Polyclonal | |
| Purified | |
| RUO | |
| NP_036281 | |
| 26581 | |
| Synthetic peptide directed towards the C terminal of human DUX5. Peptide sequence IAAREELARETGLPESRIQIWFQNRRARHRGQSGRAPTQASIRCNAAPIG. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| double homeobox 5, double homeobox protein 1, double homeobox protein 5, double homeobox, 5, DUX1 | |
| DUX5 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto