missing translation for 'onlineSavingsMsg'
Learn More

DUX4/DUX4c Antibody (2143C) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30763266 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763266 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763266 Supplier Novus Biologicals Supplier No. FAB95351JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DUX4/DUX4c Monoclonal antibody specifically detects DUX4/DUX4c in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUX4/DUX4c
Applications Western Blot
Classification Monoclonal
Clone 2143C
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias DUX10, DUX4, DUX4L
Host Species Rabbit
Immunogen Human DUX4 synthetic peptide containing sequences within amino acids 200-300 of the human DUX4 protein, Accession # Q9UBX2
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.