missing translation for 'onlineSavingsMsg'
Learn More

DUX4 Antibody (P4H2), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18422318 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422318 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422318 Supplier Novus Biologicals Supplier No. NBP149552JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DUX4 Monoclonal antibody specifically detects DUX4 in Human samples. It is validated for Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUX4
Applications Western Blot, Simple Western, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone P4H2
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias double homeobox 4, double homeobox protein 10, double homeobox protein 4, double homeobox protein 4/10, double homeobox protein DUX10, DUX10
Host Species Mouse
Immunogen Synthetic peptide corresponding to the C-terminus of human DUX4. [Swiss-Prot: Q9UBX2]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 100288687
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.