missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571298
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 77%; Human: 100%.
The human genome contains hundreds of repeats of the 3.3-kb family in regions associated with heterochromatin. The DUX gene family, including DUX1, resides within these 3.3-kb repeated elements.
Specifications
| DUX1 | |
| Polyclonal | |
| Unconjugated | |
| DUX1 | |
| double homeobox 1; double homeobox protein 1; Double homeobox protein 5; double homeobox, 1; DUX1; DUX5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26584 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O43812 | |
| DUX1 | |
| synthetic peptide directed towards the middle region of human DUX1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction