missing translation for 'onlineSavingsMsg'
Learn More

Duvelisib, MedChemExpress

Product Code. 16487288
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487288 10 mg 10mg
16477246 100 mg 100mg
16453307 50 mg 50mg
16429807 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16487288 Supplier Medchem Express Supplier No. HY17044
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Duvelisib (IPI-145) is a selectivite p100δ inhibitor with IC50 of 2.5 nM, 27.4 nM, 85 nM and 1602 nM for p110δ, P110γ, p110β and p110α, respectively.

Duvelisib (IPI-145) is a selectivite p100δ inhibitor with IC50 of 2.5 nM, 27.4 nM, 85 nM and 1602 nM for p110δ, P110γ, p110β and p110α, respectively.

Specifications

Chemical Name or Material Duvelisib
Physical Form Solid
CAS 1201438-56-3
Color Light Gray
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H17ClN6O
For Use With (Application) Cancer-Kinase/protease
Synonym IPI-145 INK1197
Solubility Information DMSO : ≥ 41 mg/mL (98.35 mM)
SMILES O=C1N(C2=CC=CC=C2)C([C@@H](NC3=C4N=CNC4=NC=N3)C)=CC5=C1C(Cl)=CC=C5
Molecular Weight (g/mol) 416.86
Formula Weight 416.86
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.