missing translation for 'onlineSavingsMsg'
Learn More

dUTPase Antibody, Novus Biologicals™

Product Code. 30231101 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30231101 100 μL 100µL
30228394 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30231101 Supplier Novus Biologicals Supplier No. NBP333441100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

dUTPase Monoclonal antibody specifically detects dUTPase in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen dUTPase
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL
Formulation PBS (pH 7.3), 50% glycerol, 0.05% BSA
Gene Alias deoxyuridine triphosphatase, dUTP pyrophosphatasedUTP nucleotidohydrolase, dUTPasedeoxyuridine 5'-triphosphate nucleotidohydrolase, mitochondrial, EC 3.6.1.23, FLJ20622
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-252 of human dUTPase (NP_001020419.1).,, Sequence:, IALPSGCYGRVAPRSGLAAKHFIDVGAGVIDEDYRGNVGVVLFNFGKEKFEVKKGDRIAQLICERIFYPEIEEVQALDDTERGSGGFGSTGKN
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1854
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.