Learn More
3M™ Dustmaster™ Particulate Filters
Product Code. 15719017
Change view
Click to view available options
Filter Type:
P2
P2R
P3
Unit Size:
10 pairs
Each
Quantity:
1 Ea.
5/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filter Type unitSize Quantity
15719017 P2R 10 pairs -
12364659 P2 Each 1 Ea.
15729797 P3 Each 5/Pk.
15719797 P2 Each 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15719017 Supplier 3M™ Supplier No. 7000037674
Only null left
Add to basket
Add to basket
Particulate filter for use with the Dustmaster Powered Air Respirator helps protects against dust, mist and fume.
For flexible P2 R protection against solid and liquid particles, use 3M™ Particulate Filters 5925. They add protection against particulates when used with 3M™ Gas and Vapour Filters, 6000 Series (using the 3M™ Filter Retainer). Our lightweight particulate filters help you work safely in hazardous environments. They’re designed to optimise your field of vision, and they can be used with any 3M™ Gas and Vapour filters, 6000 Series and 3M™ Reusable Masks that feature a bayonet-style connection.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN143 : 2000, CE Approved
Connection Type Filter retainer
Filter Type P2R
Product Type Particulate Filter
For Use With (Application) Solid and liquid particles
For Use With (Equipment) 6000 and 7000 Series face masks
Includes Filter