missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Dust Covers
GREENER_CHOICE

Product Code. 15840402
Change view
Click to view available options
Color:
Natural
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15840402 Natural Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15840402 Supplier Thermo Scientific™ Supplier No. DCVOA

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prevent contamination and protect critical samples during transport. These Dust Covers are designed to fit all standard size VOA vial caps.

Prevent contamination and protect critical samples during transport. Thermo Scientific™ Dust Covers are designed to fit all standard size VOA vial caps. Thermo Scientific accessories complement the full range of environmental sampling containers.

  • Full cap cover for VOA vials allows for septum expansion and helps to prevent contamination
  • Designed to fit all standard size VOA vial caps
  • Pre-cleaned Dust Covers available upon request

Ordering Information:

Standard package size 500 per package; other package quantities available upon request

Recommended for:

Sample transport

TRUSTED_SUSTAINABILITY

Specifications

Description Dust Cover
Diameter (English) Outer 1.1 in.
Color Natural
Diameter (Metric) Outer 27.94 mm
Material Polyethylene
Quantity 500 Cs.
Closure Size 24-414
For Use With (Equipment) VOA Vial, VOA Vial Cap
Certifications/Compliance None
Product Type Dust Cover
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.