missing translation for 'onlineSavingsMsg'
Learn More

DUSP9 Antibody (2E3), Novus Biologicals™

Product Code. 18363788 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363788 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363788 Supplier Novus Biologicals Supplier No. H00001852M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DUSP9 Monoclonal antibody specifically detects DUSP9 in Human samples. It is validated for Western Blot, ELISA, Proximity Ligation Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP9
Applications Western Blot, ELISA, Proximity Ligation Assay
Classification Monoclonal
Clone 2E3
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001386
Gene Alias dual specificity phosphatase 9, EC 3.1.3.16, EC 3.1.3.48, MAP kinase phosphatase 4, Mitogen-activated protein kinase phosphatase 4, MKP-4dual specificity protein phosphatase 9, MKP4serine/threonine specific protein phosphatase
Host Species Mouse
Immunogen DUSP9 (NP_001386, 174 a.a. ~ 278 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AESEADRDSMSCGLDSEGATPPPVGLRASFPVQILPNLYLGSARDSANLESLAKLGIRYILNVTPNLPNFFEKNGDFHYKQIPISDHWSQNLSRFFPEAIEFIDE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1852
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.