missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP8 Recombinant Rabbit Monoclonal Antibody (HL2338)

Description
DUSP8 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates SAPK/JNK and p38, is expressed predominantly in the adult brain, heart, and skeletal muscle, is localized in the cytoplasm, and is induced by nerve growth factor and insulin. An intronless pseudogene for DUSP8 is present on chromosome 10q11.2.
Specifications
Specifications
| Antigen | DUSP8 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | HL2338 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | DUSP8 |
| Gene Accession No. | Q13202 |
| Gene Alias | 5530400B01Rik; AI593498; C11orf81; dual specificity phosphatase 8; dual specificity protein phosphatase 8; Dual specificity protein phosphatase hVH-5; Dusp8; FLJ42476; FLJ42958; H1 phosphatase, vaccinia virus homolog; Hb5; HVH-5; HVH8; M3/6; neuronal tyrosine threonine phosphatase 1; neuronal tyrosine/threonine phosphatase 1; Nttp1; OTTHUMP00000164487; serine/threonine specific protein phosphatase; VH5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?