missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109668
This item is not returnable.
View return policy
Description
Immunogen sequence: NleAGDRLPR KVNl.
The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates SAPK/JNK and p38, is expressed predominantly in the adult brain, heart, and skeletal muscle, is localized in the cytoplasm, and is induced by nerve growth factor and insulin. An intronless pseudogene for DUSP8 is present on chromosome 10q11.2.
Specifications
| DUSP8 | |
| Polyclonal | |
| Unconjugated | |
| DUSP8 | |
| 5530400B01Rik; AI593498; C11orf81; dual specificity phosphatase 8; dual specificity protein phosphatase 8; Dual specificity protein phosphatase hVH-5; Dusp8; FLJ42476; FLJ42958; H1 phosphatase, vaccinia virus homolog; Hb5; HVH-5; HVH8; M3/6; neuronal tyrosine threonine phosphatase 1; neuronal tyrosine/threonine phosphatase 1; Nttp1; OTTHUMP00000164487; serine/threonine specific protein phosphatase; VH5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18218, 1850, 361679 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.97 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O09112, Q13202 | |
| DUSP8 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human DUSP8 (NP_0044112). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction