missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP8, Goat anti-Human, Polyclonal Antibody, Abnova™
Goat Polyclonal Antibody
Brand: Abnova PAB6114.100ug
This item is not returnable.
View return policy
Description
Sequence: &asterisk;AGDRLPRKVMDAK-CSpecifications
| DUSP8 | |
| Polyclonal | |
| Unconjugated | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| C11orf81/FLJ42476/FLJ42958/HB5/HVH-5/HVH8 | |
| Goat | |
| 100 μg | |
| Primary | |
| This antibody is also expected to recognize the hypothetical human protein similar to dual specificity phosphatase 8 (XM_114902), which is virtually identical. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| ELISA (1:4000) Western blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| dual specificity phosphatase 8 | |
| DUSP8 | |
| A synthetic peptide corresponding to human DUSP8. | |
| RUO | |
| 1850 | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction