missing translation for 'onlineSavingsMsg'
Learn More

DUSP8 Antibody, Novus Biologicals™

Product Code. 18276573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18276573 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18276573 Supplier Novus Biologicals Supplier No. NBP158302

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DUSP8 Polyclonal specifically detects DUSP8 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q13202
Gene Alias C11orf81, chromosome 11 open reading frame 81, dual specificity phosphatase 8, Dual specificity protein phosphatase hVH-5, EC 3.1.3.16, EC 3.1.3.48, FLJ42476, FLJ42958, HVH8, serine/threonine specific protein phosphatase, vaccinia virus homolog, VH5
Gene Symbols DUSP8
Host Species Rabbit
Immunogen Synthetic peptides corresponding to DUSP8(dual specificity phosphatase 8) The peptide sequence was selected from the middle region of DUSP8. Peptide sequence PAPPTPPATSALQQGLRGLHLSSDRLQDTNRLKRSFSLDIKSAYAPSRRP.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1850
Test Specificity Expected identity based on immunogen sequence: Bovine: 100%; Mouse: 100%; Guinea pig: 92%; Chicken: 85%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Guinea Pig
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.