missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA522014
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, H1299, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DUSP7 is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP)kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which are associatedwith cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli.
Specifications
| DUSP7 | |
| Polyclonal | |
| Unconjugated | |
| DUSP7 | |
| AU015694; dual specificity phosphatase 6; dual specificity phosphatase 7; dual specificity protein phosphatase 7; Dual specificity protein phosphatase MKP-X; dual specificity protein phosphatase PYST2; dual-specificity phosphatase-7; DUSP7; fd19d04; hypothetical protein LOC327246; MAP kinase phosphatase; Mkpx; mkp-X; PYST2; wu:fd19d04; zgc:55957; zgc:77211; zgc:77211 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1849 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q16829 | |
| DUSP7 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human DUSP7. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction