missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38011-100ul
This item is not returnable.
View return policy
Description
DUSP4 Polyclonal antibody specifically detects DUSP4 in Human samples. It is validated for ELISA,Western Blot
Specifications
| DUSP4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| dual specificity phosphatase 4, Dual specificity protein phosphatase hVH2, EC 3.1.3.16, EC 3.1.3.48, HVH2serine/threonine specific protein phosphatase, MAP kinase phosphatase 2, Mitogen-activated protein kinase phosphatase 2, MKP-2dual specificity protein phosphatase 4, MKP2VH1 homologous phosphatase 2, TYP, VH2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-168 of human DUSP4 (NP_001385.1).,, Sequence:, MVTMEELREMDCSVLKRLMNRDENGGGAGGSGSHGTLGLPSGGKCLLLDCRPFLAHSAGYILGSVNVRCNTIVRRRAKGSVSLEQILPAEEEVRARLRSGLYSAVIVYDERSPRAESLREDSTVSLVVQALRRNAERTDICLLKGGYERFSSEYPEFCSKTKALAAIP | |
| 100 μL | |
| Cell Cycle and Replication | |
| 1846 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction