missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DUSP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103034
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total DUSP3.
Protein tyrosine phosphatases (PTPs) are a group of enzymes that remove phosphate groups from phosphorylated tyrosine residues on proteins. Together with tyrosine kinases, PTPs regulate the phosphorylation state of many important signaling molecules, such as the MAP kinase family. Recently, increasing attention has been focused on the growing family of PTPs. Like PTKs, PTPs have been implicated in cell signaling, cell growth and proliferation, and oncogenic transformation. Moreover, some PTPs can be involved in cell cycle regulation and embryogenesis. Dual specificity phosphatases (DSPs) are an emerging subclass of the protein tyrosine phosphatase (PTP) gene superfamily, which appears to be selective for dephosphorylating the critical phosphothreonine and phosphotyrosine residues. The prototypical DSP is the VH1 gene in vaccinia virus expressed in late-stage viral infection. A shallow active site pocket in VHR allows for the hydrolysis of phosphorylated serine, threonine, or tyrosine protein residues, whereas the deeper active site of protein tyrosine phosphatases (PTPs) restricts substrate specificity to only phosphotyrosine.
Specifications
| DUSP3 | |
| Polyclonal | |
| Unconjugated | |
| Dusp3 | |
| 2210015O03Rik; 5031436O03Rik; dual specificity phosphatase 3; dual specificity phosphatase 3 (vaccinia virus phosphatase VH1-related); dual specificity phosphatase 3 (vaccinia virus phosphatase VH1-related)-like; dual specificity protein phosphatase 3; Dual specificity protein phosphatase VHR; DUSP 3; DUSP3; DUSP-3; RGD1560049; serine/threonine specific protein phosphatase; T-DSP11; Vaccinia H1-related phosphatase; vaccinia virus phosphatase VH1-related; VHR | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1845, 72349 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P51452, Q9D7X3 | |
| Dusp3 | |
| A synthesized peptide derived from human DUSP3(Accession P51452), corresponding to amino acid residues M1-I48. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction