missing translation for 'onlineSavingsMsg'
Learn More

DUSP27/DUPD1 Antibody (OTI7E4), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18139884 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18139884 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18139884 Supplier Novus Biologicals Supplier No. NBP272503JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DUSP27/DUPD1 Monoclonal specifically detects DUSP27/DUPD1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP27/DUPD1
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI7E4
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias atypical dual-specific protein phosphatase, Dual specificity phosphatase 27, dual specificity phosphatase and pro isomerase domain containing 1, DUSP27dual specificity phosphatase DUPD1, EC 3.1.3.16, EC 3.1.3.48, FMDSP
Gene Symbols DUPD1
Host Species Mouse
Immunogen Full length recombinant protein of human DUSP27/DUPD1 (NP_001003892) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 338599.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.