missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP26 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31254
This item is not returnable.
View return policy
Description
DUSP26 Polyclonal specifically detects DUSP26 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| DUSP26 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| DSP-4, dual specificity phosphatase 26 (putative), Dual specificity phosphatase SKRP3, dual specificity protein phosphatase 26, dual-specificity phosphatase SKRP3, DUSP24SKRP3, EC 3.1.3.16, EC 3.1.3.48, LDP4, LDP-4, Low-molecular-mass dual-specificity phosphatase 4, MAP kinase phosphatase 8, MGC1136, MGC2627, Mitogen-activated protein kinase phosphatase 8, MKP8, MKP-8, NATA1, Novel amplified gene in thyroid anaplastic cancer | |
| Rabbit | |
| 24 kDa | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neurotransmission, Protein Phosphatase | |
| 78986 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1.01mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation | |
| Q9BV47 | |
| DUSP26 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human DUSP26. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and Mouse. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (94%) and Bovine (81%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction