missing translation for 'onlineSavingsMsg'
Learn More

DUSP26 Antibody, FITC, Novus Biologicals™

Product Code. 18348432 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348432 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18348432 Supplier Novus Biologicals Supplier No. NBP306507F

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DUSP26 Polyclonal antibody specifically detects DUSP26 in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DUSP26
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias DSP-4, dual specificity phosphatase 26 (putative), Dual specificity phosphatase SKRP3, dual specificity protein phosphatase 26, dual-specificity phosphatase SKRP3, DUSP24SKRP3, EC 3.1.3.16, EC 3.1.3.48, LDP4, LDP-4, Low-molecular-mass dual-specificity phosphatase 4, MAP kinase phosphatase 8, MGC1136, MGC2627, Mitogen-activated protein kinase phosphatase 8, MKP8, MKP-8, NATA1, Novel amplified gene in thyroid anaplastic cancer
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human DUSP26 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neurotransmission, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 78986
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.